Assay Details
| Gene Symbol: LOC101115632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973653.1 | XP_027829454.1 | 12 - 13 | 1187 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115632| Gene Symbol: LOC101115632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973653.1 | XP_027829454.1 | 12 - 13 | 1187 | 99 | |
Set Membership: |
|