Assay Details
| Gene Symbol: LOC101103695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960735.1 | XP_027816536.1 | 14 - 15 | 1735 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103695| Gene Symbol: LOC101103695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960735.1 | XP_027816536.1 | 14 - 15 | 1735 | 97 | |
Set Membership: |
|