Assay Details
| Gene Symbol: LOC105616273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967386.1 | XP_027823187.1 | 33 - 34 | 3942 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105616273| Gene Symbol: LOC105616273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967386.1 | XP_027823187.1 | 33 - 34 | 3942 | 93 | |