Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096789.2 | XP_014952275.2 | 15 - 16 | 1879 | 137 | |
| XM_015096790.2 | XP_014952276.2 | 13 - 14 | 1818 | 137 | ||
More Information
Set Membership: |
|

CERT1
Genomic Map