Assay Details
| Gene Symbol: LOC101122428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967912.1 | XP_027823713.1 | 4 - 5 | 520 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122428| Gene Symbol: LOC101122428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967912.1 | XP_027823713.1 | 4 - 5 | 520 | 89 | |
Set Membership: |
|