Assay Details
| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975402.1 | XP_027831203.1 | 6 - 7 | 1124 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTOP2| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975402.1 | XP_027831203.1 | 6 - 7 | 1124 | 64 | |
Set Membership: |
|