Assay Details
| Gene Symbol: LOC101103947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003982.4 | XP_004004031.1 | 1 - 2 | 146 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103947| Gene Symbol: LOC101103947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003982.4 | XP_004004031.1 | 1 - 2 | 146 | 84 | |