Assay Details
| Gene Symbol: LOC101106372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099717.2 | XP_011955107.2 | 4 - 5 | 5414 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106372| Gene Symbol: LOC101106372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099717.2 | XP_011955107.2 | 4 - 5 | 5414 | 54 | |
Set Membership: |
|