Assay Details
| Gene Symbol: LOC101106719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957647.1 | XP_027813448.1 | 20 - 21 | 3243 | 87 | |
| XM_027957648.1 | XP_027813449.1 | 21 - 22 | 3132 | 87 | ||
| XM_027957649.1 | XP_027813450.1 | 20 - 21 | 3243 | 87 | ||
More Information
Set Membership: |
|

LOC101106719
Genomic Map