Assay Details
| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962516.1 | XP_027818317.1 | 6 - 7 | 2822 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC110| Gene Symbol: CCDC110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962516.1 | XP_027818317.1 | 6 - 7 | 2822 | 72 | |
Set Membership: |
|