Assay Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960276.1 | XP_027816077.1 | 16 - 17 | 1989 | 63 | |
| XM_027960277.1 | XP_027816078.1 | 16 - 17 | 1986 | 63 | ||
| XM_027960278.1 | XP_027816079.1 | 16 - 17 | 1983 | 63 | ||
| XM_027960279.1 | XP_027816080.1 | 16 - 17 | 1980 | 63 | ||
| XM_027960280.1 | XP_027816081.1 | 16 - 17 | 1989 | 63 | ||
| XM_027960281.1 | XP_027816082.1 | 16 - 17 | 1986 | 63 | ||
| XM_027960282.1 | XP_027816083.1 | 16 - 17 | 1983 | 63 | ||
| XM_027960283.1 | XP_027816084.1 | 16 - 17 | 1980 | 63 | ||
| XM_027960284.1 | XP_027816085.1 | 17 - 18 | 2202 | 63 | ||
More Information
Set Membership: |
|

JAKMIP3
Genomic Map