Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956500.1 | XP_027812301.1 | 3 - 4 | 725 | 77 | |
| XM_027956501.1 | XP_027812302.1 | 3 - 4 | 632 | 77 | ||
More Information
Set Membership: |
|

FAM216A
Genomic Map