Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190277.3 | XP_012045667.2 | 3 - 4 | 1589 | 109 | |
XM_012190278.3 | XP_012045668.2 | 4 - 5 | 689 | 109 | ||
XM_027977399.1 | XP_027833200.1 | 4 - 5 | 1683 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
