Assay Details
| Gene Symbol: CAVIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101052.2 | XP_014956538.2 | 1 - 2 | 399 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAVIN3| Gene Symbol: CAVIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101052.2 | XP_014956538.2 | 1 - 2 | 399 | 57 | |
Set Membership: |
|