Assay Details
| Gene Symbol: LMNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967801.1 | XP_027823602.1 | 9 - 10 | 1265 | 82 | |
| XM_027967802.1 | XP_027823603.1 | 7 - 8 | 1110 | 82 | ||
| XM_027967803.1 | XP_027823604.1 | 7 - 8 | 867 | 82 | ||

LMNTD1
Genomic Map