Assay Details
| Gene Symbol: LOC101106520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968373.1 | XP_027824174.1 | 9 - 10 | 1190 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106520| Gene Symbol: LOC101106520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968373.1 | XP_027824174.1 | 9 - 10 | 1190 | 81 | |
Set Membership: |
|