Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001903.4 | XP_004001952.1 | 3 - 4 | 350 | 65 | |
| XM_012123215.3 | XP_011978605.1 | 3 - 4 | 350 | 65 | ||
More Information
Set Membership: |
|

KIF2C
Genomic Map