Assay Details
| Gene Symbol: LOC101112936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002644.4 | XP_004002693.2 | 4 - 5 | 749 | 76 | |
| XM_027976821.1 | XP_027832622.1 | 4 - 5 | 751 | 76 | ||
| GenBank mRNA | KX931094.1 | - | 4 - 5 | 751 | 76 | |
| KX931095.1 | - | 4 - 5 | 751 | 76 | ||
| KX931096.1 | - | 4 - 5 | 751 | 76 | ||
More Information
Set Membership: |
|

LOC101112936
Genomic Map