Assay Details
| Gene Symbol: LOC101112936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002644.4 | XP_004002693.2 | 8 - 9 | 1733 | 116 | |
| GenBank mRNA | KX931094.1 | - | 8 - 9 | 1735 | 116 | |
| KX931095.1 | - | 7 - 8 | 1318 | 116 | ||
More Information
Set Membership: |
|

LOC101112936
Genomic Map