Assay Details
| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007769.3 | XP_004007818.2 | 7 - 8 | 1115 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKMY2| Gene Symbol: ANKMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007769.3 | XP_004007818.2 | 7 - 8 | 1115 | 69 | |
Set Membership: |
|