Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008112.4 | XP_004008161.1 | 3 - 4 | 382 | 101 | |
| XM_012177248.3 | XP_012032638.1 | 3 - 4 | 407 | 101 | ||
| XM_012177249.3 | XP_012032639.1 | 2 - 3 | 312 | 101 | ||
More Information
Set Membership: |
|

MRPS33
Genomic Map