Assay Details
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971929.1 | XP_027827730.1 | 3 - 4 | 355 | 77 | |
XM_027971930.1 | XP_027827731.1 | 2 - 3 | 728 | 77 | ||
XM_027971931.1 | XP_027827732.1 | 3 - 4 | 359 | 77 | ||
XM_027971932.1 | XP_027827733.1 | 3 - 4 | 479 | 77 | ||
XM_027971933.1 | XP_027827734.1 | 3 - 4 | 451 | 77 | ||
XM_027971934.1 | XP_027827735.1 | 3 - 4 | 276 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
