Assay Details
| Gene Symbol: LOC101118372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975899.1 | XP_027831700.1 | 3 - 4 | 276 | 66 | |
| XM_027975900.1 | XP_027831701.1 | 3 - 4 | 393 | 66 | ||
More Information
Set Membership: |
|

LOC101118372
Genomic Map