Assay Details
| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022395.4 | XP_004022444.2 | 10 - 11 | 2234 | 55 | |
| XM_012107481.3 | XP_011962871.2 | 11 - 12 | 2393 | 55 | ||
More Information
Set Membership: |
|

LONRF3
Genomic Map