Assay Details
| Gene Symbol: DNAJC21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097094.3 | XP_011952484.2 | 5 - 6 | 894 | 62 | |
| XM_012097095.3 | XP_011952485.2 | 5 - 6 | 770 | 62 | ||
More Information
Set Membership: |
|

DNAJC21
Genomic Map