Assay Details
| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104797.2 | XP_011960187.2 | 3 - 4 | 641 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YPEL3| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104797.2 | XP_011960187.2 | 3 - 4 | 641 | 104 | |
Set Membership: |
|