Assay Details
| Gene Symbol: RIPPLY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012108635.3 | XP_011964025.2 | 1 - 2 | 104 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIPPLY3| Gene Symbol: RIPPLY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012108635.3 | XP_011964025.2 | 1 - 2 | 104 | 117 | |
Set Membership: |
|