Assay Details
| Gene Symbol: LOC101121185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974935.1 | XP_027830736.1 | 12 - 13 | 1787 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101121185| Gene Symbol: LOC101121185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974935.1 | XP_027830736.1 | 12 - 13 | 1787 | 78 | |
Set Membership: |
|