Assay Details
| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973464.1 | XP_027829265.1 | 2 - 3 | 216 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALKAL1| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973464.1 | XP_027829265.1 | 2 - 3 | 216 | 89 | |