Assay Details
| Gene Symbol: LOC101104748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970643.1 | XP_027826444.1 | 11 - 12 | 1813 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104748| Gene Symbol: LOC101104748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970643.1 | XP_027826444.1 | 11 - 12 | 1813 | 61 | |
Set Membership: |
|