Assay Details
| Gene Symbol: LOC101104748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970643.1 | XP_027826444.1 | 2 - 3 | 618 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104748| Gene Symbol: LOC101104748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970643.1 | XP_027826444.1 | 2 - 3 | 618 | 121 | |
Set Membership: |
|