Assay Details
| Gene Symbol: LOC101118793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972290.1 | XP_027828091.1 | 14 - 15 | 2013 | 132 | |
| XM_027972291.1 | XP_027828092.1 | 13 - 14 | 1992 | 132 | ||
| XM_027972292.1 | XP_027828093.1 | 7 - 8 | 1340 | 132 | ||
| Gene Symbol: LOC114108752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972289.1 | XP_027828090.1 | 6 - 7 | 952 | 132 | |
More Information
Set Membership: |
|

LOC101118793
Genomic Map