Assay Details
Gene Symbol: LOC101112889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104769.2 | XP_014960255.1 | 1 | 211 | 86 |
Gene Symbol: LOC114111539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963359.1 | XP_027819160.1 | 1 | 211 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
