Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003405.3 | XP_004003454.1 | 3 - 4 | 423 | 89 | |
| XM_015092705.2 | XP_014948191.1 | 3 - 4 | 775 | 89 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map