Assay Details
| Gene Symbol: LOC114116429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973289.1 | XP_027829090.1 | 1 | 189 | 82 | |
| Gene Symbol: RFFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975294.1 | XP_027831095.1 | 2 - 3 | 377 | 82 | |
More Information
Set Membership: |
|

LOC114116429
Genomic Map