Assay Details
| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012810.4 | XP_004012859.2 | 4 - 5 | 378 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDK5RAP3| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012810.4 | XP_004012859.2 | 4 - 5 | 378 | 63 | |
Set Membership: |
|