Assay Details
| Gene Symbol: LOC114116628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973704.1 | XP_027829505.1 | 1 | 95 | 73 | |
| Gene Symbol: SETD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956265.1 | XP_027812066.1 | 2 - 3 | 1404 | 73 | |
More Information
Set Membership: |
|

LOC114116628
Genomic Map