Assay Details
| Gene Symbol: NMNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975948.1 | XP_027831749.1 | 10 - 11 | 926 | 53 | |
| XM_027975949.1 | XP_027831750.1 | 9 - 10 | 811 | 53 | ||
| XM_027975950.1 | XP_027831751.1 | 9 - 10 | 805 | 53 | ||
More Information
Set Membership: |
|

NMNAT2
Genomic Map