Assay Details
Gene Symbol: C5H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177809.3 | XP_012033199.2 | 1 - 2 | 81 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012177809.3 transcripts.
Set Membership: |
![]() ![]() |
