Assay Details
| Gene Symbol: C5H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177809.3 | XP_012033199.2 | 6 - 7 | 1784 | 85 | |
| XM_027969515.1 | XP_027825316.1 | 6 - 7 | 2037 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H19orf57| Gene Symbol: C5H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177809.3 | XP_012033199.2 | 6 - 7 | 1784 | 85 | |
| XM_027969515.1 | XP_027825316.1 | 6 - 7 | 2037 | 85 | ||