Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 18 - 19 | 2120 | 82 | |
| XM_027969087.1 | XP_027824888.1 | 21 - 22 | 2577 | 82 | ||
| XM_027969088.1 | XP_027824889.1 | 18 - 19 | 2379 | 82 | ||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map