Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 20 - 21 | 2352 | 77 | |
| XM_027969087.1 | XP_027824888.1 | 23 - 24 | 2809 | 77 | ||
| XM_027969088.1 | XP_027824889.1 | 20 - 21 | 2611 | 77 | ||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map