Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 3 - 4 | 526 | 76 | |
| XM_027969087.1 | XP_027824888.1 | 5 - 6 | 917 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027969086.1 transcripts.
Set Membership: |
|

DYNC2I1
Genomic Map