Assay Details
| Gene Symbol: RACGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006369.4 | XP_004006418.1 | 16 - 17 | 1907 | 56 | |
| XM_012174327.3 | XP_012029717.1 | 16 - 17 | 1903 | 56 | ||
| XM_012174328.3 | XP_012029718.1 | 16 - 17 | 1900 | 56 | ||
More Information
Set Membership: |
|

RACGAP1
Genomic Map