Assay Details
| Gene Symbol: ATP5F1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008775.4 | XP_004008824.1 | 3 - 4 | 671 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5F1D| Gene Symbol: ATP5F1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008775.4 | XP_004008824.1 | 3 - 4 | 671 | 83 | |
Set Membership: |
|