Assay Details
| Gene Symbol: ABHD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018415.3 | XP_004018464.1 | 3 - 4 | 542 | 80 | |
| XM_012099976.3 | XP_011955366.2 | 2 - 3 | 958 | 80 | ||
| XM_027957573.1 | XP_027813374.1 | 3 - 4 | 1522 | 80 | ||
More Information
Set Membership: |
|

ABHD14A
Genomic Map