Assay Details
| Gene Symbol: ORAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104494.3 | XP_011959884.1 | 3 - 4 | 3139 | 109 | |
| XM_012104495.3 | XP_011959885.1 | 2 - 3 | 3320 | 109 | ||
| XM_012104496.3 | XP_011959886.1 | 3 - 4 | 2046 | 109 | ||
| XM_012104497.3 | XP_011959887.1 | 2 - 3 | 3011 | 109 | ||
| XM_027961221.1 | XP_027817022.1 | 3 - 4 | 3980 | 109 | ||
| XM_027961222.1 | XP_027817023.1 | 3 - 4 | 1283 | 109 | ||
| XM_027961223.1 | XP_027817024.1 | 3 - 4 | 2050 | 109 | ||
| XM_027961225.1 | XP_027817026.1 | 2 - 3 | 315 | 109 | ||
More Information
Set Membership: |
|

ORAI2
Genomic Map