Assay Details
| Gene Symbol: UBE2L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979791.1 | XP_027835592.1 | 3 - 4 | 377 | 80 | |
| XM_027979792.1 | XP_027835593.1 | 2 - 3 | 253 | 80 | ||
More Information
Set Membership: |
|

UBE2L6
Genomic Map