Assay Details
| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098558.3 | XP_011953948.1 | 1 - 2 | 616 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL2A1| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098558.3 | XP_011953948.1 | 1 - 2 | 616 | 99 | |
Set Membership: |
|