Assay Details
| Gene Symbol: SLC7A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958409.1 | XP_027814210.1 | 7 - 8 | 2337 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC7A14| Gene Symbol: SLC7A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958409.1 | XP_027814210.1 | 7 - 8 | 2337 | 67 | |
Set Membership: |
|